Learn More
Teknova LB Agar Plates with Ampicillin-100 and Chloramphenicol-34. 100mm, 20 Plates per Sleeve, Sterile. 8 pack.
SDP
Catalog No. 502387527
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,100g/mL Ampicillin Sodium Salt,34g/mL Chloramphenicol
Catalog No. 50-238-7527 Supplier Teknova Supplier No. L12538PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,100g/mL Ampicillin Sodium Salt,34g/mL Chloramphenicol