Learn More
Teknova LB Agar Plates with Ampicillin-100 and Kanamycin-50. 100mm, 20 plates/sleeve, Sterile. 8 pack.
SDP
Catalog No. 502387867
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,100g/mL Ampicillin Sodium Salt,50g/mL Kanamycin Sulfate
Catalog No. 50-238-7867 Supplier Teknova Supplier No. L12108PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,100g/mL Ampicillin Sodium Salt,50g/mL Kanamycin Sulfate