Learn More
Teknova LB Agar Plates with Ampicillin-100. 100mm, 20 Plates per Sleeve, Sterile.
SDP
Catalog No. 502388036
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 100¿g/mL Ampicillin Sodium Salt
Catalog No. 50-238-8036 Supplier Teknova Supplier No. L1004
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 100¿g/mL Ampicillin Sodium Salt