Learn More
Teknova LB Agar Plates with Ampicillin-100. 86x128mm, 20 Plates per Sleeve, Sterile. 8 pack.
SDP
Catalog No. 502387898
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,100g/mL Ampicillin Sodium Salt
Catalog No. 50-238-7898 Supplier Teknova Supplier No. L20048PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,100g/mL Ampicillin Sodium Salt