Learn More
Teknova LB Agar Plates with Ampicillin-100. 100mm, 35mL Fill, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387312
Encompass
1.0% Tryptone, 0.5% Tastone 900, 1.0% Sodium Chloride, 1.8% Agar,100g/mL Ampicillin Sodium Salt
Catalog No. 50-238-7312 Supplier Teknova Supplier No. 2L10054PK
1.0% Tryptone, 0.5% Tastone 900, 1.0% Sodium Chloride, 1.8% Agar,100g/mL Ampicillin Sodium Salt