Learn More
Teknova LB Agar Plates with Ampicillin-100. 100X100mm, 20 Plates per Sleeve, Sterile. 8 pack.
SDP
Catalog No. 502387341
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,100g/ml Ampicillin
Catalog No. 50-238-7341 Supplier Teknova Supplier No. L30048PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,100g/ml Ampicillin