Learn More
Teknova LB Agar Plates with Ampicillin-100 and Chloramphenicol-25. 100mm, 20 Plates/Sleeve, Sterile. 8 pack.
SDP
Catalog No. 502387371
Encompass
1% Tryptone, 0.5% Yeast extract, 1.0% Sodium chloride, 1.5% Agar, 100g/mL Ampicillin Sodium Salt, 25g/mL Chloramphenicol
Catalog No. 50-238-7371 Supplier Teknova Supplier No. L12468PK
1% Tryptone, 0.5% Yeast extract, 1.0% Sodium chloride, 1.5% Agar, 100g/mL Ampicillin Sodium Salt, 25g/mL Chloramphenicol