Learn More
Teknova LB Agar Plates with Ampicillin-50, X-Gal, IPTG. 100mm, 20 Plates per Sleeve, Sterile. 8 pack.
SDP
Supplier:  Teknova L19008PK
Encompass
1% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 50g/mL Ampicillin Sodium Salt, 60g/mL X-Gal, 0.1mM Isopropyl-B-D-thiogalactopyranoside(IPTG)
Catalog No. 50-238-7358