Learn More
Teknova LB Agar Plates with Animal-Free Soytone & Tet-12.5. 100mm, 20 Plates per Sleeve, Sterile 8 pack.
SDP
Catalog No. 502387260
Encompass
1.69% Animal-Free Soytone, 0.50% Yeast Extract,1.00% Sodium Chloride, 1.50% Agar,12.5g/mL Tetracycline Hydrochloride
Catalog No. 50-238-7260 Supplier Teknova Supplier No. L15678PK
1.69% Animal-Free Soytone, 0.50% Yeast Extract,1.00% Sodium Chloride, 1.50% Agar,12.5g/mL Tetracycline Hydrochloride