Learn More
Teknova LB Agar Plates with Animal Free Soytone and Chloramphenicol-34. 100mm, 20 Plates per Sleeve, Sterile 4 pack.
SDP
Catalog No. 502387868
Encompass
1.69% Animal-Free Soytone, 0.50% Yeast Extract,1.00% Sodium Chloride, 1.50% Agar, 34g/mL Chloramphenicol
Catalog No. 50-238-7868 Supplier Teknova Supplier No. L15704PK
1.69% Animal-Free Soytone, 0.50% Yeast Extract,1.00% Sodium Chloride, 1.50% Agar, 34g/mL Chloramphenicol