Learn More
Teknova LB Agar Plates with Animal-Free Soytone and Chloramphenicol-25. 100mm, 20 Plates per Sleeve, Sterile 4 Pack.
SDP
Catalog No. 502387366
Encompass
1.7% Animal-Free Soytone, 0.5% Yeast Extract,1.0% Sodium Chloride, 1.5% Agar,25g/mL Chloramphenicol
Catalog No. 50-238-7366 Supplier Teknova Supplier No. L15164PK
1.7% Animal-Free Soytone, 0.5% Yeast Extract,1.0% Sodium Chloride, 1.5% Agar,25g/mL Chloramphenicol