Learn More
Teknova LB Agar Plates with Animal-Free Soytone and Kanamycin-30. 100mm, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Supplier:  Teknova L11244PK
Encompass
1.69% Animal-Free Soytone, 0.50% Yeast Extract, 1.00% Sodium Chloride, 1.50% Agar, 30g/mL Kanamycin Sulfate
Catalog No. 50-238-7767