Learn More
Teknova LB Agar Plates with Animal-Free Soytone and Kanamycin-50. 100mm, 20 Plates per Sleeve, Sterile.
SDP
Catalog No. 502388152
Encompass
1.69% Animal-Free Soytone, 0.50% Yeast Extract,1.00% Sodium Chloride, 1.50% Agar, 50g/mL Kanamycin Sulfate
Catalog No. 50-238-8152 Supplier Teknova Supplier No. L1096
1.69% Animal-Free Soytone, 0.50% Yeast Extract,1.00% Sodium Chloride, 1.50% Agar, 50g/mL Kanamycin Sulfate