Learn More
Teknova LB Agar Plates with Animal-Free Soytone and Carbenicillin-100. 100mm, 20 Plates/Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502388004
Encompass
1.7% Animal-Free Soytone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 100g/mL Carbenicillin Disodium Salt
Catalog No. 50-238-8004 Supplier Teknova Supplier No. L10924PK
1.7% Animal-Free Soytone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 100g/mL Carbenicillin Disodium Salt