Learn More
Teknova LB Agar Plates with Animal-Free Soytone. 100mm, 20 Plates per Sleeve, Sterile.
SDP
Catalog No. 502388118
Encompass
1.7% Animal-Free Soytone, 0.5% Yeast Extract,1.0% Sodium Chloride, 1.5% Agar
Catalog No. 50-238-8118 Supplier Teknova Supplier No. L1000
1.7% Animal-Free Soytone, 0.5% Yeast Extract,1.0% Sodium Chloride, 1.5% Agar