Learn More
Teknova LB Agar Plates with Animal-Free Soytone & Kan-100 . 100mm, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387265
Encompass
1.69% Soytone, Animal-Free, 0.50% Yeast Extract, 1.00% Sodium Chloride, 1.50% Agar, 100g/mL Kanamycin Sulfate
Catalog No. 50-238-7265 Supplier Teknova Supplier No. L10974PK
1.69% Soytone, Animal-Free, 0.50% Yeast Extract, 1.00% Sodium Chloride, 1.50% Agar, 100g/mL Kanamycin Sulfate