Learn More
Teknova LB Agar Plates with Animal-Free Soytone and Carbenicillin-50. 100mm, 20 Plates per Sleeve, Sterile. 4 Pack.
SDP
Catalog No. 502387503
Encompass
1.7% Soytone, Animal-free, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 50g/mL Carbenicillin Disodium Salt
Catalog No. 50-238-7503 Supplier Teknova Supplier No. L11184PK
1.7% Soytone, Animal-free, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 50g/mL Carbenicillin Disodium Salt