Learn More
Teknova LB Agar Plates with Animal-Free Soytone and Ampicillin-100. 100mm, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387479
Encompass
1.0% Animal-Free Soytone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 100g/mL Ampicillin Sodium Salt
Catalog No. 50-238-7479 Supplier Teknova Supplier No. L10934PK
1.0% Animal-Free Soytone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 100g/mL Ampicillin Sodium Salt