Learn More
Teknova LB Agar Plates with Carbenicillin-100. 150mm, 10 Plates per Sleeve, Sterile. 8 pack.
SDP
Catalog No. 502387654
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,100g/mL Carbenicillin Disodium Salt
Catalog No. 50-238-7654 Supplier Teknova Supplier No. L11418PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,100g/mL Carbenicillin Disodium Salt