Learn More
Teknova LB Agar Plates with Carbenicillin-100. 100mm, 10 Plates per Sleeve, Sterile. 8 Pack.
SDP
Catalog No. 502387863
Encompass
1.0% Tryptone, 0.5% Yeast extract, 1.0% Sodium chloride, 1.5% Agar,100g/mL Carbenicillin Disodium Salt
Catalog No. 50-238-7863 Supplier Teknova Supplier No. L51408PK
1.0% Tryptone, 0.5% Yeast extract, 1.0% Sodium chloride, 1.5% Agar,100g/mL Carbenicillin Disodium Salt