Learn More
Teknova LB Agar Plates with Carbenicillin-100, 60µg/mL X-Gal, 0.3mM IPTG. 100mm Plates, 20 Plates per Sleeve, Sterile. 4 Pack.
SDP
Catalog No. 502387363
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 100g/mL Carbenicillin Disodium Salt, 60g/mL X-Gal, 0.3mM Isopropyl-B-D-thiogalactopyranoside(IPTG)
Catalog No. 50-238-7363 Supplier Teknova Supplier No. L19634PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 100g/mL Carbenicillin Disodium Salt, 60g/mL X-Gal, 0.3mM Isopropyl-B-D-thiogalactopyranoside(IPTG)