Learn More
Teknova LB Agar Plates with Carb-100, 8 Well, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502388012
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,100g/mL Carbenicillin Disodium Salt
Catalog No. 50-238-8012 Supplier Teknova Supplier No. L24184PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,100g/mL Carbenicillin Disodium Salt