Learn More
Teknova LB Agar Plates with Carb-100 and 1% Glucose. 150mm, 20 Plates per Sleeve, Sterile.[H0] 8 pack.
SDP
Catalog No. 502387520
Encompass
1.0% Tryptone, 0.5% Yeast extract, 1.0% Sodium chloride, 1.5% Agar, 1.0% Cerelose Anhydrous Dextrose (Glucose), 100g/mL Carbenicillin
Catalog No. 50-238-7520 Supplier Teknova Supplier No. L58018PK
1.0% Tryptone, 0.5% Yeast extract, 1.0% Sodium chloride, 1.5% Agar, 1.0% Cerelose Anhydrous Dextrose (Glucose), 100g/mL Carbenicillin