Learn More
Teknova LB Agar Plates with Carbenicillin-100, X-Gal & IPTG. 100mm, 10 Plates per Sleeve, Sterile. 8 Pack.
SDP
Supplier:  Teknova L51428PK
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,100g/mL Carbenicillin Disodium Salt, 60g/mL X-Gal, 0.1mM Isopropyl-B-D-thiogalactopyranoside(IPTG)
Catalog No. 50-238-7641