Learn More
Teknova LB Agar Plates with Carbenicillin-100. 245mm, 200mL Fill, 4 Plates per Sleeve, Sterile. 4 Pack.
SDP
Catalog No. 502387386
Encompass
1.0% Tryptone, 0.5% Yeast extract, 1.0% Sodium chloride, 1.5% Agar,100g/mL Carbenicillin Disodium Salt
Catalog No. 50-238-7386 Supplier Teknova Supplier No. L61664PK
1.0% Tryptone, 0.5% Yeast extract, 1.0% Sodium chloride, 1.5% Agar,100g/mL Carbenicillin Disodium Salt