Learn More
Teknova LB Agar Plates with Carbenicillin-50. 100mm, 20 Plates per Sleeve, Sterile. (Heat Shrink) 8 Pack.
SDP
Catalog No. 502387624
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,50g/mL Carbenicillin Disodium Salt
Catalog No. 50-238-7624 Supplier Teknova Supplier No. L70088PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,50g/mL Carbenicillin Disodium Salt