Learn More
Teknova LB Agar Plates with Carb-50. 100X100mm, 20 Plates per Sleeve, Sterile. 8 pack.
SDP
Catalog No. 502387435
Encompass
1.0% Tryptone, 0.5% Yeast extract, 1.0% Sodium chloride, 1.5% Agar, 50g/mL Carbenicillin Disodium Salt
Catalog No. 50-238-7435 Supplier Teknova Supplier No. L30088PK
1.0% Tryptone, 0.5% Yeast extract, 1.0% Sodium chloride, 1.5% Agar, 50g/mL Carbenicillin Disodium Salt