Learn More
Teknova LB Agar Plates with Carbenicillin-150. 100mm, 20 Plates per Sleeve, Sterile. 8 pack.
SDP
Supplier:  Teknova L10118PK
Encompass
1.00% Tryptone, 0.50% Yeast Extract, 1.00% Sodium Chloride, 1.50% Agar,150g/mL Carbenicillin Disodium Salt
Catalog No. 50-238-7849