Learn More
Teknova LB Agar Plates with Carbenicillin-100. 100mm, 20 Plates per Sleeve, Sterile. (Heat Shrink)
SDP
Catalog No. 502387978
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,100g/mL Carbenicillin Disodium Salt
Catalog No. 50-238-7978 Supplier Teknova Supplier No. L7010
Add to Cart
Edge
Add to Cart
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,100g/mL Carbenicillin Disodium Salt