Learn More
Teknova LB Agar Plates with Carbenicillin-50. 100mm, 20 Plates per Sleeve, Sterile.
SDP
Catalog No. 502387998
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,50g/mL Carbenicillin Disodium Salt
Catalog No. 50-238-7998 Supplier Teknova Supplier No. L1008
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,50g/mL Carbenicillin Disodium Salt