Learn More
Teknova LB Agar Plates with Carbenicillin-100. 86x128mm, 20 Plates per Sleeve, Sterile. 4 Pack.
SDP
Catalog No. 502388150
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,100g/mL Carbenicillin Disodium Salt
Catalog No. 50-238-8150 Supplier Teknova Supplier No. L20104PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,100g/mL Carbenicillin Disodium Salt