Learn More
Teknova LB Agar Plates with 1% Glucose and Carbenicillin-100. 100mm, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502388035
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 1.0% Glucose, 100g/mL Carbenicillin Disodium Salt
Catalog No. 50-238-8035 Supplier Teknova Supplier No. L18014PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 1.0% Glucose, 100g/mL Carbenicillin Disodium Salt