Learn More
Teknova LB Agar Plates with Carbenicillin-100. 245mm Corning Low Profile, 4 Plates/Sleeve, Sterile.
SDP
Catalog No. 502388047
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,100g/mL Carbenicillin Disodium Salt
Catalog No. 50-238-8047 Supplier Teknova Supplier No. L6010
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,100g/mL Carbenicillin Disodium Salt