Learn More
Teknova LB Agar Plates with Carbenicillin-100 and Chloramphenicol-25. 100mm, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502388054
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 100g/mL Carbenicillin Disodium Salt, 25g/mL Chloramphenicol
Catalog No. 50-238-8054 Supplier Teknova Supplier No. L13174PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 100g/mL Carbenicillin Disodium Salt, 25g/mL Chloramphenicol