Learn More
Teknova LB Agar Plates with Carbenicillin-100. 100mm, 20 Plates per Sleeve, Sterile.
SDP
Catalog No. 502388149
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,100¿g/mL Carbenicillin Disodium Salt
Catalog No. 50-238-8149 Supplier Teknova Supplier No. L1010
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,100¿g/mL Carbenicillin Disodium Salt