Learn More
Teknova LB Agar Plates with Carbenicillin-200. 150mm, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387535
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,200g/mL Carbenicillin Disodium Salt
Catalog No. 50-238-7535 Supplier Teknova Supplier No. L50464PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,200g/mL Carbenicillin Disodium Salt