Learn More
Teknova LB Agar Plates with Carbenicillin-100 and Kanamycin-50. 100mm, 20 Plates/Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387326
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,100g/mL Carbenicillin Disodium Salt, 50g/mL Kanamycin Sulfate
Catalog No. 50-238-7326 Supplier Teknova Supplier No. L12884PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,100g/mL Carbenicillin Disodium Salt, 50g/mL Kanamycin Sulfate