Learn More
Teknova LB Agar Plates with Carbenicillin-100 and Chloramphenicol-12.5. 100mm, 20 Plates/Sleeve, Sterile. 8 pack.
SDP
Catalog No. 502387496
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 100.0g/mL Carbenicillin Disodium Salt, 12.5g/mL Chloramphenicol
Catalog No. 50-238-7496 Supplier Teknova Supplier No. L12838PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 100.0g/mL Carbenicillin Disodium Salt, 12.5g/mL Chloramphenicol