Learn More
Teknova LB Agar Plates with Chloramphenicol-50. 100mm, 20 Plates per Sleeve, Sterile.
SDP
Catalog No. 502387995
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,50g/mL Chloramphenicol
Catalog No. 50-238-7995 Supplier Teknova Supplier No. L1019
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,50g/mL Chloramphenicol