Learn More
Teknova LB Agar Plates with Chloramphenicol-34. 100mm, 20 Plates per Sleeve, Sterile.
SDP
Catalog No. 502387997
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,34g/mL Chloramphenicol
Catalog No. 50-238-7997 Supplier Teknova Supplier No. L1017
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,34g/mL Chloramphenicol