Learn More
Teknova LB Agar Plates with Chloramphenicol-34 and Kanamycin-50. 100mm, 20 Plates per Sleeve, Sterile.
SDP
Catalog No. 502388124
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,34g/mL Chloramphenicol, 50g/mL Kanamycin Sulfate
Catalog No. 50-238-8124 Supplier Teknova Supplier No. L1232
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,34g/mL Chloramphenicol, 50g/mL Kanamycin Sulfate