Learn More
Teknova LB Agar Plates with Chloramphenicol-20. 100mm, 20 Plates per Sleeve, Sterile. 8 pack.
SDP
Catalog No. 502387410
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,20g/mL Chloramphenicol
Catalog No. 50-238-7410 Supplier Teknova Supplier No. L10158PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,20g/mL Chloramphenicol