Learn More
Teknova LB Agar Plates with Chloramphenicol-25 and Kanamycin-50. 100mm, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387629
Encompass
1.0% Tryptone, 0.5% Yeast Extract,1.0% Sodium Chloride, 1.5% Agar,25g/mL Chloramphenicol, 50g/mL Kanamycin Sulfate
Catalog No. 50-238-7629 Supplier Teknova Supplier No. L12304PK
1.0% Tryptone, 0.5% Yeast Extract,1.0% Sodium Chloride, 1.5% Agar,25g/mL Chloramphenicol, 50g/mL Kanamycin Sulfate