Learn More
Teknova LB Agar Plates with Chloramphenicol-30. 100mm, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387584
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,30g/mL Chloramphenicol
Catalog No. 50-238-7584 Supplier Teknova Supplier No. L19974PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,30g/mL Chloramphenicol