Learn More
Teknova LB Agar Plates with Chloramphenicol-12.5. 150mm, 20 Plates per Sleeve, Sterile. 8 pack.
SDP
Catalog No. 502387598
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,12.5g/mL Chloramphenicol
Catalog No. 50-238-7598 Supplier Teknova Supplier No. L50138PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,12.5g/mL Chloramphenicol