Learn More
Teknova LB Agar Plates with Chloramphenicol-12.5 and Kanamycin-50. 100mm, 20 Plates/Sleeve, Sterile. 8 pack.
SDP
Catalog No. 502387906
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 12.5g/mL Chloramphenicol, 50.0g/mL Kanamycin Sulfate
Catalog No. 50-238-7906 Supplier Teknova Supplier No. L12588PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 12.5g/mL Chloramphenicol, 50.0g/mL Kanamycin Sulfate