Learn More
Teknova LB Agar Plates with Chloramphenicol-12.5. 100mm, 20 Plates per Sleeve, Sterile.
SDP
Catalog No. 502388198
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,12.5g/mL Chloramphenicol
Catalog No. 50-238-8198 Supplier Teknova Supplier No. L1013
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,12.5g/mL Chloramphenicol