Learn More
Teknova LB Agar Plates with Chloramphenicol-25. 100mm, 20 Plates per Sleeve, Sterile
SDP
Catalog No. 502387993
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 25g/mL Chloramphenicol
*Item is refrigerated.
Catalog No. 50-238-7993 Supplier Teknova Supplier No. L1016
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 25g/mL Chloramphenicol
*Item is refrigerated.