Learn More
Teknova LB Agar Plates with Chloramphenicol-5. 100mm, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Supplier:  Teknova L10414PK
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,5g/mL Chloramphenicol
Catalog No. 50-238-7445