Learn More
Teknova LB Agar Plates with Gentamicin-7, Kanamycin-50, Tetracyline-10, Bluo-gal, IPTG. 100mm, 20 Plates per Sleeve. 8 Pack.
SDP
Catalog No. 502387585
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,50g/mL Kanamycin Sulfate, 7g/mL Gentamicin Sulfate, 10g/mL Tetracycline Hydrochloride, 150g/mL Bluo-Gal, 40g/mL Isopropyl-B-D-thiogalactopyranoside(IPTG)
Catalog No. 50-238-7585 Supplier Teknova Supplier No. L19248PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,50g/mL Kanamycin Sulfate, 7g/mL Gentamicin Sulfate, 10g/mL Tetracycline Hydrochloride, 150g/mL Bluo-Gal, 40g/mL Isopropyl-B-D-thiogalactopyranoside(IPTG)